Learn More
Alkali Scientific D Pantothenic Acid Hemicalcium Salt, 100 G
SDP
Catalog No. 5000661129
Encompass_Preferred
D Pantothenic Acid Hemicalcium Salt is a stable, water-soluble form of vitamin B5 commonly used in biochemical research. This 100-gram bottle is ideal for larger-scale applications in studies involving metabolism, coenzyme A synthesis, and energy production in cells. As a precursor for coenzyme A, pantothenic acid plays a critical role in cellular processes such as fatty acid metabolism, acetylation, and energy production. It is particularly useful in studies of cellular bioenergetics, enzymatic reactions, and nutritional biochemistry. Its widespread use in metabolic and pharmaceutical research makes it a key reagent in many laboratory settings.
Catalog No. 50-006-61129 Supplier Alkali Scientific Supplier No. DPA003CP
D Pantothenic Acid Hemicalcium Salt is a stable, water-soluble form of vitamin B5 commonly used in biochemical research. This 100-gram bottle is ideal for larger-scale applications in studies involving metabolism, coenzyme A synthesis, and energy production in cells. As a precursor for coenzyme A, pantothenic acid plays a critical role in cellular processes such as fatty acid metabolism, acetylation, and energy production. It is particularly useful in studies of cellular bioenergetics, enzymatic reactions, and nutritional biochemistry. Its widespread use in metabolic and pharmaceutical research makes it a key reagent in many laboratory settings.