Learn More
Medchemexpress LLC D-Pantothenic acid hemicalcium salt | 137-08-6 | 238.27 | 50 G
SDP
Catalog No. 5000416789
Encompass_Preferred
D-Pantothenic acid hemicalcium salt is an orally active vitamin that reduces patulin in apple juice. It removes reactive oxygen species (ROS) and has antibacterial and immunomodulatory activities. It also has effects on improving osteoporosis and obesity.
  • Restrains Th17 cell differentiation through CoA in a PANK-dependent manner.
  • Inhibits RANKL induced osteoclastogenesis by scavenging ROS generation in BMMs cells.
  • Rescues a membrane trafficking defect in human fibroblasts devoid of TANGO2.
  • Protects against obesity via brown adipose tissue activation.
  • Alleviates experimental autoimmune encephalomyelitis.
  • Alleviates bone loss in an osteoporotic model of OVX mice.
  • Reduces bacterial growth via regulating innate immunity and adaptive immunity.
  • Enhances intestinal development and alters microbes in weaned piglets.
Catalog No. 50-004-16789 Supplier Medchemexpress LLC Supplier No. HYN068150G
Add to Cart
Add to Cart
D-Pantothenic acid hemicalcium salt is an orally active vitamin that reduces patulin in apple juice. It removes reactive oxygen species (ROS) and has antibacterial and immunomodulatory activities. It also has effects on improving osteoporosis and obesity.
  • Restrains Th17 cell differentiation through CoA in a PANK-dependent manner.
  • Inhibits RANKL induced osteoclastogenesis by scavenging ROS generation in BMMs cells.
  • Rescues a membrane trafficking defect in human fibroblasts devoid of TANGO2.
  • Protects against obesity via brown adipose tissue activation.
  • Alleviates experimental autoimmune encephalomyelitis.
  • Alleviates bone loss in an osteoporotic model of OVX mice.
  • Reduces bacterial growth via regulating innate immunity and adaptive immunity.
  • Enhances intestinal development and alters microbes in weaned piglets.