Learn More
STEMCELL Technologies D-PBS, 10X Concentrate (Without Ca++ and Mg++), Size: 500 mL
SDP
Catalog No. 502390222
Encompass
Dulbecco’s phosphate-buffered saline (D-PBS) can be used as a temporary diluting washing irrigating or transporting solution for cell or tissue culture. It provides a buffering system to maintain the physiological pH range and osmotic balance of culture media and provides cells with a source of water and essential inorganic ions.
Catalog No. 50-239-0222 Supplier STEMCELL Technologies Supplier No. 37354
Add to Cart
Add to Cart
Dulbecco’s phosphate-buffered saline (D-PBS) can be used as a temporary diluting washing irrigating or transporting solution for cell or tissue culture. It provides a buffering system to maintain the physiological pH range and osmotic balance of culture media and provides cells with a source of water and essential inorganic ions.