Learn More
Electron Microscopy Sciences D-PBS, Formula I Phosphate Buffer Saline 500 ML
SDP
Catalog No. 50980259
Encompass_Preferred
All of our Dullbecco's Phosphate Buffer Saline Solutions contain Potassium Chloride, Potassium Phosphate Monobasic, Sodium Chloride, Sodium Phosphate Dibasic, and D-Glucose. Some formulations may contain additional components. Solutions are prepared 1X strength.
Catalog No. 50-980-259 Supplier Electron Microscopy Sciences Supplier No. 1177205
All of our Dullbecco's Phosphate Buffer Saline Solutions contain Potassium Chloride, Potassium Phosphate Monobasic, Sodium Chloride, Sodium Phosphate Dibasic, and D-Glucose. Some formulations may contain additional components. Solutions are prepared 1X strength.