Learn More
Electron Microscopy Sciences D-PBS, Formula II Phosphate Buffer Saline 1 LT
SDP
Catalog No. 50980262
Encompass_Preferred
All of our Dullbecco's Phosphate Buffer Saline Solutions contain Potassium Chloride, Potassium Phosphate Monobasic, Sodium Chloride, Sodium Phosphate Dibasic, and D-Glucose. Some formulations may contain additional components. Solutions are prepared 1X strength.
Catalog No. 50-980-262 Supplier Electron Microscopy Sciences Supplier No. 1177310
All of our Dullbecco's Phosphate Buffer Saline Solutions contain Potassium Chloride, Potassium Phosphate Monobasic, Sodium Chloride, Sodium Phosphate Dibasic, and D-Glucose. Some formulations may contain additional components. Solutions are prepared 1X strength.