Learn More
STEMCELL Technologies D-PBS (Without Ca++ and Mg++), Size: 500 mL
SDP
Catalog No. NC1128272
Encompass
Dulbecco’s phosphate-buffered saline (D-PBS) can be used as a temporary diluting, washing, irrigating, or transporting solution for cell or tissue culture. It provides a buffering system to maintain the physiological pH range and osmotic balance of culture media, and provides cells with a source of water and essential inorganic ions.
Catalog No. NC1128272 Supplier STEMCELL Technologies Supplier No. 37350
Dulbecco’s phosphate-buffered saline (D-PBS) can be used as a temporary diluting, washing, irrigating, or transporting solution for cell or tissue culture. It provides a buffering system to maintain the physiological pH range and osmotic balance of culture media, and provides cells with a source of water and essential inorganic ions.