Learn More
Apexbio Technology LLC D-Penicillamine, 100mg CAS# 52-67-5
SDP
Supplier:  Apexbio Technology LLC B8392100
Encompass_Preferred
Penicillamine is an exogenous NOS regulator and an anti-rheumatic and chelating agent for Wilson's disease. Used as anti-rheumatic drugs and chelating agents in hepatolenticular degeneration. Used as a copper chelating agent to form mixed disulfides with cysteine or other sulfide media components. Used for inactivation of protein-1 DNA binding, inhibiting the growth of rabbit articular chondrocytes in asynchronous culture. Other sizes are also available. Please inqury us for quote.
Catalog No. 50-199-8432