Learn More
D-(-)-Penicillamine, 98%
Description
It is used mainly as chelating agent in heavy metal poisoning as in lead, mercury and copper poisoning. It is used in Wilson's disease. It is used in pharmacy as chelating agent and as immunosuppressant for treating rheumatoid arthritis by reducing the number of T cells, inhibiting the macrophages, by reducing the activity of Interleukin 1 and rheumatoid factor and in preventing crosslinking of the collagen. It acts as an exogenous NOS modulator.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
It is freely soluble in water, slightly soluble in alcohol, insoluble in chloroform, ether, acetone, benzene, and in carbon tetrachloride.
Notes
Stable under normal conditions. Incompatabile to strong oxidizing agents. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Recommended storage temperature: 2 - 8°C.
Specifications
Specifications
| Melting Point | 202°C to 204°C (decomposition) |
| pH | 4.5 to 5.5 |
| Odor | Unpleasant |
| Quantity | 25 g |
| Beilstein | 1722375 |
| Merck Index | 14,7088 |
| Solubility Information | It is freely soluble in water,slightly soluble in alcohol,insoluble in chloroform,ether,acetone,benzene,and in carbon tetrachloride. |
| Optical Rotation | −63° (c=1 in 1N NaOH) |
| Formula Weight | 149.21 |
| Percent Purity | 99% |
| Show More |
Safety and Handling
| Hazard Category | H315-H319-H334 |
| Hazard Statement | GHS H Statement H315-H319-H335 Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
| Precautionary Statement | P261-P264b-P280-P285-P302+P352-P304+P341-P305+P351+P338-P332+P313-P342+P311-P362-P501c |
| EINECSNumber | 200-148-8 |
| RTECSNumber | YV9425000 |
| TSCA | No |
| Recommended Storage | Keep cold |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.