Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

D-Penicillamine, MP Biomedicals™

Catalog No. MP215180505 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP215180505 5 g
MP215180501 1 g
MP215180525 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. MP215180505 Supplier MP Biomedicals, Inc Supplier No. 0215180505
Add to Cart
Add to Cart

Used as a physiological chelating agent for heavy metals. D-Penicillamine, MP Biomedicals is a characteristic degradation product of penicillin type antibiotics. It interferes with the formation of cross-links between tropocollagen molecules and lowers Ig

Penicillamine combines one atom of copper with two molecules of penicillamine. It reduces excess cystine excretion in cystinuria by disulfide interchange between penicillamine and cysteine. This results in formation of a readily excreted penicillamine-cysteine disulfide. Penicillamine interferes with the formation of cross-links between tropocollagen molecules and cleaves them when newly formed. Penicillamine lowers IgM rheumatoid factor and depresses T-cell activity.

  • Used as an antirheumatic and a chelating agent in Wilson′s disease
  • Used as a copper chelator to form mixed disulfides with cysteine or other sulfide media components
  • Used to inactivate protein-1 DNA binding
  • Used to inhibit the growth of asynchronous cultures of rabbit articular chondrocytes
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 52-67-5
Molecular Formula C5H11NO2S
Molecular Weight (g/mol) 149.208
InChI Key VVNCNSJFMMFHPL-VKHMYHEASA-N
Synonym d-penicillamine, penicillamine, cuprimine, d---penicillamine, depen, 3-mercapto-d-valine, cuprenil, d-penamine, --penicillamine, d-mercaptovaline
PubChem CID 5852
ChEBI CHEBI:7959
IUPAC Name (2S)-2-amino-3-methyl-3-sulfanylbutanoic acid
SMILES CC(C)(C(C(=O)O)N)S

Specifications

Melting Point 210°C (decomposition)(literature)
Quantity 5 g
Solubility Information Soluble in water (100mg/mL).
Formula Weight 149.2
Percent Purity ≥99%
Physical Form Powder
Recommended Storage Store at 4°C.
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.