Learn More
Medchemexpress LLC D-Phenyl-alanine-N-t-Boc-d5 | 1213661-19-8 | 99.9% | 270.34 | 10 MG
SDP
Catalog No. 5000366296
Encompass_Preferred
D-Phenyl-alanine-N-t-Boc-d5 is the deuterium labeled D-Phenyl-alanine-N-t-Boc. It is intended for research use only.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Stable heavy isotopes have been incorporated into drug molecules, largely as tracers for quantitation during the drug development process.
  • Deuteration has the potential to affect the pharmacokinetic and metabolic profiles of drugs.
Catalog No. 50-003-66296 Supplier Medchemexpress LLC Supplier No. HYW009412S10MG
D-Phenyl-alanine-N-t-Boc-d5 is the deuterium labeled D-Phenyl-alanine-N-t-Boc. It is intended for research use only.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Stable heavy isotopes have been incorporated into drug molecules, largely as tracers for quantitation during the drug development process.
  • Deuteration has the potential to affect the pharmacokinetic and metabolic profiles of drugs.