Learn More
Medchemexpress LLC HY-Y0079 100mg Medchemexpress, D-Phenylalanine CAS:673-06-3 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYY0079100MG
Encompass_Preferred
Medchemexpress, HY-Y0079 100mg D-Phenylalanine CAS:673-06-3 D-Phenylalanine is the synthetic dextro isomer of phenylalanine. D-Phenylalanine inhibits biofilm development of Pseudoalteromonas sp. SC2014 [1] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-4673