Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

D-phenylalanine, 99.9%, MP Biomedicals™

Catalog No. ICN10261605 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
5 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN10261605 5 g
ICN10261680 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. ICN10261605 Supplier MP Biomedicals, Inc Supplier No. 0210261605

Chemical Identifiers

CAS 673-06-3
Molecular Formula C9H11NO2
Molecular Weight (g/mol) 165.19
MDL Number MFCD00004270
InChI Key COLNVLDHVKWLRT-MRVPVSSYSA-N
Synonym d-phenylalanine, h-d-phe-oh, 2r-2-amino-3-phenylpropanoic acid, d-phe, sabiden, phenylalanine d-form, alanine, phenyl-, d, d-diphenylalanine, r-2-amino-3-phenylpropanoic acid, d-alpha-amino-beta-phenylpropionic acid
PubChem CID 71567
ChEBI CHEBI:16998
IUPAC Name (2R)-2-amino-3-phenylpropanoic acid
SMILES N[C@H](CC1=CC=CC=C1)C(O)=O

Specifications

Color White
Quantity 5 g
Formula Weight 165.2
Percent Purity 99.9%
Physical Form Powder
Chemical Name or Material D-Phenylalanine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.