Learn More
Medchemexpress LLC D-phenylalanine-d5 | 362049-55-6 | MFCD01861577 | 99.9% | 170.22 | C9H6D5NO2 | 5mg
SDP
Catalog No. 5000269034
Encompass_Preferred
D-Phenylalanine-d5 is the deuterium labeled D-Phenylalanine D-Phenylalanine is the synthetic dextro isomer of phenylalanine D-Phenylalanine inhibits biofilm development of Pseudoalteromonas sp SC2014[1]
Catalog No. 50-002-69034 Supplier Medchemexpress LLC Supplier No. HYY0079S5MG
D-Phenylalanine-d5 is the deuterium labeled D-Phenylalanine D-Phenylalanine is the synthetic dextro isomer of phenylalanine D-Phenylalanine inhibits biofilm development of Pseudoalteromonas sp SC2014[1]