Learn More
Medchemexpress LLC D-phenylalanine-d8 | 1202064-02-5 | 98.3% | 173.24 g/mol | C9H3D8NO2 | 5mg
SDP
Catalog No. 5000269165
Encompass_Preferred
D-Phenylalanine-d8 is the deuterium labeled D-Phenylalanine D-Phenylalanine is the synthetic dextro isomer of phenylalanine D-Phenylalanine inhibits biofilm development of Pseudoalteromonas sp SC2014[1]
Catalog No. 50-002-69165 Supplier Medchemexpress LLC Supplier No. HYY0079S15MG
D-Phenylalanine-d8 is the deuterium labeled D-Phenylalanine D-Phenylalanine is the synthetic dextro isomer of phenylalanine D-Phenylalanine inhibits biofilm development of Pseudoalteromonas sp SC2014[1]