Learn More
Medchemexpress LLC D-(-)-α-phenylglycine ethyl ester hydrochloride | 17609-48-2 | MFCD00052005 | 99.3% | 215.68 | C10H14ClNO2 | 500 G
SDP
Catalog No. 5000303859
Encompass_Preferred
D-(-)-α-Phenylglycine ethyl ester hydrochloride is an optically active glycine derivative supplied as the hydrochloride salt. It is used as a chiral building block and intermediate in peptide synthesis and asymmetric organic synthesis.
  • Optically active chiral building block for peptide and asymmetric synthesis.
  • High purity (reported >99%) suitable for analytical and synthetic applications.
  • White to off-white solid, easy to handle and weigh.
  • Recommended sealed storage at 4°C to maintain stability.
  • Available in multiple pack sizes up to 500 g for scale-up.
Catalog No. 50-003-03859 Supplier Medchemexpress LLC Supplier No. HYW014168500G
D-(-)-α-Phenylglycine ethyl ester hydrochloride is an optically active glycine derivative supplied as the hydrochloride salt. It is used as a chiral building block and intermediate in peptide synthesis and asymmetric organic synthesis.
  • Optically active chiral building block for peptide and asymmetric synthesis.
  • High purity (reported >99%) suitable for analytical and synthetic applications.
  • White to off-white solid, easy to handle and weigh.
  • Recommended sealed storage at 4°C to maintain stability.
  • Available in multiple pack sizes up to 500 g for scale-up.