Learn More
Medchemexpress LLC HY-30219 100mg Medchemexpress, D-​(+)​-​Phenyllactic acid CAS:7326-19-4 Purity:>98%
SDP
Catalog No. 501932851
Encompass_Preferred
Medchemexpress, HY-30219 100mg D-​(+)​-​Phenyllactic acid CAS:7326-19-4 D-​(+)​-​Phenyllactic acid is an anti-bacterial agent, excreted by Geotrichum candidum, inhibits a range of Gram-positive from humans and foodstuffs and Gram-negative bacteria found in humans [1] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2851 Supplier Medchemexpress LLC Supplier No. HY30219100MG
Medchemexpress, HY-30219 100mg D-​(+)​-​Phenyllactic acid CAS:7326-19-4 D-​(+)​-​Phenyllactic acid is an anti-bacterial agent, excreted by Geotrichum candidum, inhibits a range of Gram-positive from humans and foodstuffs and Gram-negative bacteria found in humans [1] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.