Learn More
Apexbio Technology LLC D-Pinitol 10284-63-6 20mg
SDP
Catalog No. 5000090126
Encompass_Preferred
D-Pinitol (CAS 10284-63-6) is a small-molecule insulin-mimetic compound targeting glucose transporter GLUT4 It is designed to enhance glucose uptake and modulate glucose metabolism thereby improving insulin signaling pathways D-Pinitol exerts its biological activity primarily by increasing GLUT4 translocation and modulating phosphatidylinositol 3-kinase (PI3K) and protein kinase B (Akt) activation It demonstrates inhibitory activity against various viral strains with reported IC50 values in the micromolar range in vitro Based on these pharmacological properties D-Pinitol holds research potential in insulin resistance diabetic complications and cardiovascular protective mechanisms
Catalog No. 50-000-90126 Supplier Apexbio Technology LLC Supplier No. N230720
D-Pinitol (CAS 10284-63-6) is a small-molecule insulin-mimetic compound targeting glucose transporter GLUT4 It is designed to enhance glucose uptake and modulate glucose metabolism thereby improving insulin signaling pathways D-Pinitol exerts its biological activity primarily by increasing GLUT4 translocation and modulating phosphatidylinositol 3-kinase (PI3K) and protein kinase B (Akt) activation It demonstrates inhibitory activity against various viral strains with reported IC50 values in the micromolar range in vitro Based on these pharmacological properties D-Pinitol holds research potential in insulin resistance diabetic complications and cardiovascular protective mechanisms