Learn More
Medchemexpress LLC (D)-PPA 1 TFA | 99.3% | 1669.67 | 5 MG
SDP
Catalog No. 5000399452
Encompass_Preferred
(D)-PPA 1 TFA is a hydrolysis-resistant d-peptide antagonist that serves as a potent PD-1/PD-L1 inhibitor. It binds to PD-1 with an affinity of 0.51 μM, demonstrating both in vitro and in vivo efficacy.
  • Hydrolysis-resistant d-peptide antagonist
  • Potent PD-1/PD-L1 inhibitor
  • Binds to PD-1 with an affinity of 0.51 μM
  • Blocks the interaction between PD-1/PD-L1
  • Inhibits CT26 tumor growth in mice
  • Targets tumor tissue in CT26-tumor-bearing mice
Catalog No. 50-003-99452 Supplier Medchemexpress LLC Supplier No. HYP4072A10MG
(D)-PPA 1 TFA is a hydrolysis-resistant d-peptide antagonist that serves as a potent PD-1/PD-L1 inhibitor. It binds to PD-1 with an affinity of 0.51 μM, demonstrating both in vitro and in vivo efficacy.
  • Hydrolysis-resistant d-peptide antagonist
  • Potent PD-1/PD-L1 inhibitor
  • Binds to PD-1 with an affinity of 0.51 μM
  • Blocks the interaction between PD-1/PD-L1
  • Inhibits CT26 tumor growth in mice
  • Targets tumor tissue in CT26-tumor-bearing mice