Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC D-praziquanamine ((-)-praziquanamine) | 55375-92-3 | MFCD30491837 | 98.9% | 202.26 g/mol | C12H14N2O | 10 MG
SDP

Catalog No. 5000297861
Encompass_Preferred

D-Praziquanamine ((-)-praziquanamine) is the stereoisomer of L-praziquanamine, provided as a research reagent for use as an experimental control in biochemical and pharmacological studies.

  • Isomer of L-praziquanamine, intended for use as an experimental control.
  • CAS number 55375-92-3.
  • Molecular formula C12H14N2O, molecular weight 202.26 g/mol.
  • Typical purity ~98.9% (batch-specific; see certificate of analysis for details).
  • Appearance: solid; also offered as a 10 mM solution in DMSO.
  • Available in small sample sizes and milligram-scale quantities (for example, 10 mg).

Catalog No. 50-002-97861 Supplier Medchemexpress LLC Supplier No. HY4165610MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

D-Praziquanamine ((-)-praziquanamine) is the stereoisomer of L-praziquanamine, provided as a research reagent for use as an experimental control in biochemical and pharmacological studies.

  • Isomer of L-praziquanamine, intended for use as an experimental control.
  • CAS number 55375-92-3.
  • Molecular formula C12H14N2O, molecular weight 202.26 g/mol.
  • Typical purity ~98.9% (batch-specific; see certificate of analysis for details).
  • Appearance: solid; also offered as a 10 mM solution in DMSO.
  • Available in small sample sizes and milligram-scale quantities (for example, 10 mg).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.