Learn More
Medchemexpress LLC D-Pro-Phe-Arg-Chloromethylketone | 88546-74-1 | 99.5% | 450.96 | 1 MG
SDP
Supplier:  Medchemexpress LLC HYP42451MG
Encompass_Preferred
This product is a chemical compound acting as an inhibitor of coagulation factor XII and plasma kallikrein. It has a significant role in research related to thrombosis and inflammation. The compound is provided as a white to off-white solid with a purity of 99.51% and is intended for research use only, not for human use.
  • Inhibits coagulation factor XII.
  • Inhibits plasma kallikrein.
  • Plays an important role in thrombosis studies.
  • Plays an important role in inflammation studies.
  • Provided as a white to off-white solid.
  • Suitable for in vitro research.
  • High purity of 99.51%.
Catalog No. 50-003-99595