Learn More
Medchemexpress LLC [D-Pro2] Spantide I TFA | 99.2% | 10 MG
SDP
Catalog No. 5000423634
Encompass_Preferred
[D-Pro2] Spantide I TFA is a Spantide analog that acts as a selective NK1 receptor antagonist. This product is intended for research use only.
  • Selective NK1 receptor antagonist
  • Shipped at room temperature (continental US, may vary elsewhere)
  • Store as powder at -80°C for 2 years or -20°C for 1 year
  • Store in solvent at -80°C for 6 months or -20°C for 1 month
  • Recommended for research use only
Catalog No. 50-004-23634 Supplier Medchemexpress LLC Supplier No. HYP1194B10MG
[D-Pro2] Spantide I TFA is a Spantide analog that acts as a selective NK1 receptor antagonist. This product is intended for research use only.
  • Selective NK1 receptor antagonist
  • Shipped at room temperature (continental US, may vary elsewhere)
  • Store as powder at -80°C for 2 years or -20°C for 1 year
  • Store in solvent at -80°C for 6 months or -20°C for 1 month
  • Recommended for research use only