Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

D-(-)-Prolinamide, 98%

Catalog No. aah3090103
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3090103 1 g
AAH3090106 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH3090103 Supplier Thermo Scientific Chemicals Supplier No. H3090103
Only null left
Add to Cart

CAS: 62937-45-5 | C5H10N2O | 114.15 g/mol

It can be used in the synthesis of proline derivatives as efficient organocatalysts for aldol reaction on water.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It can be used in the synthesis of proline derivatives as efficient organocatalysts for aldol reaction on water.

Solubility
Soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 62937-45-5
Molecular Formula C5H10N2O
Molecular Weight (g/mol) 114.15
MDL Number MFCD00153457
InChI Key VLJNHYLEOZPXFW-UHFFFAOYNA-N
Synonym d-prolinamide, r-prolinamide, h-d-pro-nh2, r-pyrrolidine-2-carboxamide, 2r-pyrrolidine-2-carboxamide, d-proline amide, h-d-pro-nh, d-pro-nh2, 2r-2-carbamoylpyrrolidine, 2-pyrrolidinecarboxamide, 2r
PubChem CID 447554
IUPAC Name (2R)-pyrrolidine-2-carboxamide
SMILES NC(=O)C1CCCN1

Specifications

Melting Point 100°C to 103°C
Quantity 1 g
Solubility Information Soluble in water.
Optical Rotation +107° (c=2 in Ethanol)
Formula Weight 114.15
Percent Purity 98%
Chemical Name or Material D-(-)-Prolinamide
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.