Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

D-Proline, ≥99%, MP Biomedicals™

Catalog No. ICN19545605 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN19545605 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN19545605 Supplier MP Biomedicals, Inc Supplier No. 0219545605
Add to Cart
Add to Cart

Chemical Identifiers

CAS 344-25-2
Molecular Formula C5H9NO2
Molecular Weight (g/mol) 115.132
MDL Number MFCD00064317
InChI Key ONIBWKKTOPOVIA-SCSAIBSYSA-N
Synonym d-proline, 2r-pyrrolidine-2-carboxylic acid, r-pyrrolidine-2-carboxylic acid, h-d-pro-oh, d-pro, d-+-proline, d-prolin, proline, r-proline, d +-proline
PubChem CID 8988
ChEBI CHEBI:16313
IUPAC Name (2R)-pyrrolidine-2-carboxylic acid
SMILES C1CC(NC1)C(=O)O

Specifications

Color White
Assay Percent Range ≥99%
Quantity 5 g
Formula Weight 115.1
Physical Form Powder
Chemical Name or Material D-Proline
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.