Learn More
Sigma Aldrich Fine Chemicals Biosciences D Psicose 95100MG
Catalog No. 502902591
Encompass
D-Psicose protects and preserves pancreatic -islets through the maintenance of hyperglycemia and by the prevention of fat accumulation in Otsuka Long-Evans Tokushima fatty rats.
Catalog No. 50-290-2591 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. P8043100MG
D-Psicose protects and preserves pancreatic -islets through the maintenance of hyperglycemia and by the prevention of fat accumulation in Otsuka Long-Evans Tokushima fatty rats.