Learn More
Medchemexpress LLC D-Psicose-d | 53685-24-8 | 99.9% | 181.16 | 25 MG
SDP
Catalog No. 5000426139
Encompass_Preferred
D-Psicose-d is a deuterium-labeled D-Psicose, an orally active rare sugar. It inhibits p38-MAPK phosphorylation and MCP-1 expression, and also inhibits the AGEs/RAGE/NF-κB pathway. It protects pancreatic β-islets, and improves hyperglycemia and high-fat diet-induced non-alcoholic fatty liver disease.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Deuteration can affect pharmacokinetic and metabolic profiles of drugs
  • For research use only
Catalog No. 50-004-26139 Supplier Medchemexpress LLC Supplier No. 25MGHYN9914S
D-Psicose-d is a deuterium-labeled D-Psicose, an orally active rare sugar. It inhibits p38-MAPK phosphorylation and MCP-1 expression, and also inhibits the AGEs/RAGE/NF-κB pathway. It protects pancreatic β-islets, and improves hyperglycemia and high-fat diet-induced non-alcoholic fatty liver disease.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Deuteration can affect pharmacokinetic and metabolic profiles of drugs
  • For research use only