Learn More
Medchemexpress LLC D-Psicose-d (D-Allulose-d) | 53685-24-8 | 99.9% | 181.16 | 10 MG
SDP
Catalog No. 5000426369
Encompass_Preferred
D-Psicose-d is a deuterium-labeled form of D-Psicose, which is an orally active rare sugar. D-psicose has been shown to inhibit p38-MAPK phosphorylation and MCP-1 expression, as well as the AGEs/RAGE/NF-κB pathway. It also protects pancreatic β-islets and improves hyperglycemia and non-alcoholic fatty liver disease induced by a high-fat diet. This compound can be utilized as a tracer or an internal standard for quantitative analysis methods such as NMR, GC-MS, or LC-MS. Deuteration, the incorporation of stable heavy isotopes into drug molecules, is used for quantitation during drug development and has the potential to influence the pharmacokinetic and metabolic profiles of drugs.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
Catalog No. 50-004-26369 Supplier Medchemexpress LLC Supplier No. HYN9914S10MG
D-Psicose-d is a deuterium-labeled form of D-Psicose, which is an orally active rare sugar. D-psicose has been shown to inhibit p38-MAPK phosphorylation and MCP-1 expression, as well as the AGEs/RAGE/NF-κB pathway. It also protects pancreatic β-islets and improves hyperglycemia and non-alcoholic fatty liver disease induced by a high-fat diet. This compound can be utilized as a tracer or an internal standard for quantitative analysis methods such as NMR, GC-MS, or LC-MS. Deuteration, the incorporation of stable heavy isotopes into drug molecules, is used for quantitation during drug development and has the potential to influence the pharmacokinetic and metabolic profiles of drugs.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.