Learn More
Medchemexpress LLC D-Psicose (Standard) | 551-68-8 | MFCD00083478 | 99.9% | 180.16 | 10 MG
SDP
Catalog No. 5000426742
Encompass_Preferred
D-Psicose (Standard) is the analytical standard of D-Psicose, an orally active rare sugar. It is intended for research and analytical applications, commonly used in qualitative, quantitative, and methodological research experiments in HPLC, GC, and MS.
  • Analytical standard for D-Psicose
  • Intended for research and analytical applications
  • Orally active rare sugar
  • Inhibits p38-MAPK phosphorylation and MCP-1 expression
  • Inhibits the AGEs/RAGE/NF-κB pathway
  • Protects pancreatic β-islets
  • Improves hyperglycemia
  • Improves high-fat diet-induced non-alcoholic fatty liver disease
  • Used in HPLC, GC, and MS for qualitative, quantitative, and methodological research
Catalog No. 50-004-26742 Supplier Medchemexpress LLC Supplier No. HYN9914R10MG
D-Psicose (Standard) is the analytical standard of D-Psicose, an orally active rare sugar. It is intended for research and analytical applications, commonly used in qualitative, quantitative, and methodological research experiments in HPLC, GC, and MS.
  • Analytical standard for D-Psicose
  • Intended for research and analytical applications
  • Orally active rare sugar
  • Inhibits p38-MAPK phosphorylation and MCP-1 expression
  • Inhibits the AGEs/RAGE/NF-κB pathway
  • Protects pancreatic β-islets
  • Improves hyperglycemia
  • Improves high-fat diet-induced non-alcoholic fatty liver disease
  • Used in HPLC, GC, and MS for qualitative, quantitative, and methodological research