Learn More
Cayman Chemical D-Pyroglutamic Acid, Purity: ≥98%, CAS: 4042-36-8, 50 g
SDP
Catalog No. 502988605
Encompass_Preferred
A metabolite of D-glutamate; urine levels are increased in patients with nascent metabolic syndrome; plasma levels are increased in patients with end-stage renal disease
Catalog No. 50-298-8605 Supplier Cayman Chemical Supplier No. 3117050 G
A metabolite of D-glutamate; urine levels are increased in patients with nascent metabolic syndrome; plasma levels are increased in patients with end-stage renal disease