Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

D-Pyroglutamic Acid 98.0+%, TCI America™
SDP

Catalog No. P135425G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P135425G 25 g
P13545G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P135425G Supplier TCI America Supplier No. P135425G
Only null left
Add to Cart
Add to Cart

Chemical Identifiers

CAS 4042-36-8
Molecular Formula C5H7NO3
Molecular Weight (g/mol) 129.115
MDL Number MFCD00066212
InChI Key ODHCTXKNWHHXJC-GSVOUGTGSA-N
Synonym d-pyroglutamic acid, 5-oxo-d-proline, r-5-oxopyrrolidine-2-carboxylic acid, d-pyr-oh, 2r-5-oxopyrrolidine-2-carboxylic acid, d-proline, 5-oxo, r-+-2-pyrrolidone-5-carboxylic acid, d-5-pyrrolidone-2-carboxylic acid, r-2-pyrrolidone-5-carboxylic acid, d-5-oxo-2-pyrrolidinecarboxylic acid
PubChem CID 439685
ChEBI CHEBI:16924
IUPAC Name (2R)-5-oxopyrrolidine-2-carboxylic acid
SMILES C1CC(=O)NC1C(=O)O

Specifications

Melting Point 162°C
Color White
Quantity 25 g
Formula Weight 129.12
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material D-Pyroglutamic Acid
EINECSNumber (5)-0115
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.