Learn More
Cayman Chemical D-PyrohomoglutamIc AcId 100mg
SDP
Catalog No. 502991484
Encompass_Preferred
A building block; has been used in the synthesis of LMP2 inhibitors and is a component of substance P peptide analogs
Catalog No. 50-299-1484 Supplier Cayman Chemical Supplier No. 31693100 MG
A building block; has been used in the synthesis of LMP2 inhibitors and is a component of substance P peptide analogs