Learn More
Cayman Chemical D-PyrohomoglutamIc AcId 500mg
SDP
Catalog No. 502988035
Encompass_Preferred
A building block; has been used in the synthesis of LMP2 inhibitors and is a component of substance P peptide analogs
Catalog No. 50-298-8035 Supplier Cayman Chemical Supplier No. 31693500 MG
A building block; has been used in the synthesis of LMP2 inhibitors and is a component of substance P peptide analogs