Learn More
Cayman Chemical D-Pyrohomoglutamic Acid, Purity: ≥95%, CAS: 72002-30-3, 50 mg
SDP
Catalog No. 502991485
Encompass_Preferred
A building block; has been used in the synthesis of LMP2 inhibitors and is a component of substance P peptide analogs
Catalog No. 50-299-1485 Supplier Cayman Chemical Supplier No. 3169350 MG
A building block; has been used in the synthesis of LMP2 inhibitors and is a component of substance P peptide analogs