Learn More
Medchemexpress LLC HY-N0464 100mg Medchemexpress, D-(-)-Quinic acid CAS:77-95-2 Purity:>98%
SDP
Catalog No. 501933581
Encompass_Preferred
Medchemexpress, HY-N0464 100mg D-(-)-Quinic acid CAS:77-95-2 D-(-)-Quinic acid is a cyclohexanecarboxylic acid and is implicated in the perceived acidity of coffee. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-3581 Supplier Medchemexpress LLC Supplier No. HYN0464100MG
Medchemexpress, HY-N0464 100mg D-(-)-Quinic acid CAS:77-95-2 D-(-)-Quinic acid is a cyclohexanecarboxylic acid and is implicated in the perceived acidity of coffee. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.