Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Meso Scale Discovery MSD Blocker D-R, 1 mL, for Research Use Only
SDP

Catalog No. NC2311883
Encompass

The MESO SCALE DISCOVERY Blocker D-R is a specialized blocking reagent designed for research applications. It is essential for enhancing specificity and minimizing non-specific binding in various immunoassays, which in turn improves assay performance and data reliability. This product is provided in a 1 mL volume and is strictly for research use only, not intended for diagnostic procedures.

  • Specialized blocker for immunoassays
  • Optimizes assay performance
  • Reduces non-specific binding
  • Available in a 1 mL volume
  • Research use only
  • Not intended for diagnostic use
  • Storage condition of ≤-10 °C
  • Made-to-order availability
  • Usually ships in 1-3 days

Catalog No. NC2311883 Supplier Meso Scale Discovery Supplier No. R93BR3
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

The MESO SCALE DISCOVERY Blocker D-R is a specialized blocking reagent designed for research applications. It is essential for enhancing specificity and minimizing non-specific binding in various immunoassays, which in turn improves assay performance and data reliability. This product is provided in a 1 mL volume and is strictly for research use only, not intended for diagnostic procedures.

  • Specialized blocker for immunoassays
  • Optimizes assay performance
  • Reduces non-specific binding
  • Available in a 1 mL volume
  • Research use only
  • Not intended for diagnostic use
  • Storage condition of ≤-10 °C
  • Made-to-order availability
  • Usually ships in 1-3 days

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.