Learn More
AdipoGen D-(+)-Raffinose pentahydrate (100 g)
SDP
Catalog No. 502054629
Encompass_Preferred
Chemical. CAS: 17629-30-0. Formula: C18H32O16 . 5H2O. MW: 594.51. D-(+)-Raffinose is a trisaccharide composed of galactose, glucose, and fructose that occurs naturally in a variety of vegetables and grains. It is hydrolyzed to galactose and sucrose by alpha-galactosidase. Shown to have impacts on the immune system.
Catalog No. 50-205-4629 Supplier AdipoGen Supplier No. CDXR0082G100
Chemical. CAS: 17629-30-0. Formula: C18H32O16 . 5H2O. MW: 594.51. D-(+)-Raffinose is a trisaccharide composed of galactose, glucose, and fructose that occurs naturally in a variety of vegetables and grains. It is hydrolyzed to galactose and sucrose by alpha-galactosidase. Shown to have impacts on the immune system.