Learn More
Medchemexpress LLC D(+)-Raffinose pentahydrate | 17629-30-0 | 594.51 | 5 G
SDP
Catalog No. 5000415208
Encompass_Preferred
D(+)-Raffinose pentahydrate is an orally active inhibitor of LecA (Kd = 32 μM) and GtfC. It functions by reducing cyclic diguanylate (c-di-GMP) through increased activity of a c-di-GMP specific phosphodiesterase and interferes with GTF function. This compound decreases IL-4 and IL-5 mRNA, exhibits biofilm-inhibitory activity against *Pseudomonas aeruginosa* and *Streptococcus mutans*, and inhibits allergic airway eosinophilia. It is intended for research use only and not for sale to patients.
  • Orally active inhibitor of LecA and GtfC
  • Reduces cyclic diguanylate (c-di-GMP)
  • Interferes with GTF function
  • Decreases IL-4 and IL-5 mRNA
  • Exhibits biofilm-inhibitory activity against *Pseudomonas aeruginosa* and *Streptococcus mutans*
  • Inhibits allergic airway eosinophilia
Catalog No. 50-004-15208 Supplier Medchemexpress LLC Supplier No. HYN19385G
D(+)-Raffinose pentahydrate is an orally active inhibitor of LecA (Kd = 32 μM) and GtfC. It functions by reducing cyclic diguanylate (c-di-GMP) through increased activity of a c-di-GMP specific phosphodiesterase and interferes with GTF function. This compound decreases IL-4 and IL-5 mRNA, exhibits biofilm-inhibitory activity against *Pseudomonas aeruginosa* and *Streptococcus mutans*, and inhibits allergic airway eosinophilia. It is intended for research use only and not for sale to patients.
  • Orally active inhibitor of LecA and GtfC
  • Reduces cyclic diguanylate (c-di-GMP)
  • Interferes with GTF function
  • Decreases IL-4 and IL-5 mRNA
  • Exhibits biofilm-inhibitory activity against *Pseudomonas aeruginosa* and *Streptococcus mutans*
  • Inhibits allergic airway eosinophilia