Learn More
Sigma Aldrich Fine Chemicals Biosciences D-(+)-Raffinose pentahydrate, 17629-30-0, MFCD00071590, 100G
Supplier:  Sigma Aldrich Fine Chemicals Biosciences 83400100G
Encompass
Molecular Formula: C18H32O16 · 5H2O, Molecular Weight: 594.51, Beilstein Registry Number: 99541 Assay: ≥99.0%, Solubility: H2O: 0.1 g/mL, clear, colorless
Catalog No. NC1508617