Learn More
Gold Biotechnology Inc D Raffinose pentahydrate 500 g
SDP
Catalog No. 5000562107
Encompass_Preferred
Raffinose is a natural trisaccharide composed of galactose glucose and fructose It primarily serves as a carbon source for yeast as well as being an important component in some cryopreservation solutions
Catalog No. 50-005-62107 Supplier Gold Biotechnology Inc Supplier No. R030500
Raffinose is a natural trisaccharide composed of galactose glucose and fructose It primarily serves as a carbon source for yeast as well as being an important component in some cryopreservation solutions