Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

D-(+)-Raffinose pentahydrate, 98%, ≥98%, MP Biomedicals™

Catalog No. ICN10279725 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN10279725 25 g
ICN10279780 100 g
ICN10279790 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. ICN10279725 Supplier MP Biomedicals, Inc Supplier No. 0210279725
Add to Cart
Add to Cart

Chemical Identifiers

CAS 17629-30-0
Molecular Formula C18H42O21
Molecular Weight (g/mol) 594.513
InChI Key BITMAWRCWSHCRW-ORBWWOJFSA-N
Synonym d-+-raffinosepentahydrate
PubChem CID 134129414
IUPAC Name (3S,4S,5R,6R)-2-[[(2R,3S,4S,5S)-6-[(2S,3S,4R,5R)-3,4-dihydroxy-2,5-bis(hydroxymethyl)oxolan-2-yl]oxy-3,4,5-trihydroxyoxan-2-yl]methoxy]-6-(hydroxymethyl)oxane-3,4,5-triol;pentahydrate
SMILES C(C1C(C(C(C(O1)OCC2C(C(C(C(O2)OC3(C(C(C(O3)CO)O)O)CO)O)O)O)O)O)O)O.O.O.O.O.O

Specifications

Color White
Density 1.465g/mL
Assay Percent Range 0.98
Quantity 25 g
Formula Weight 594.5
Percent Purity ≥98%
Physical Form Crystalline Powder
Chemical Name or Material D-(+)-Raffinose pentahydrate, For HPLC analysis
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.