Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

D-(+)-Raffinose pentahydrate, ≥99% (HPLC), MilliporeSigma™ Supelco™

Catalog No. 111013359 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-3359 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-3359 Supplier MilliporeSigma Supelco Supplier No. 95068100MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Raffinose pentahydrate is a crystalline form of pharmaceutical sugar, raffinose which may act as ′scavengers′ or ′reservoirs′ for water with the pharmaceutical formulations.

Featured Industries: Food and Beverages
TRUSTED_SUSTAINABILITY

Specifications

CAS 17629-30-0
Linear Formula C18H32O16 · 5H2 O
MDL Number MFCD00071590
Synonym O-alpha-D-Galactopyranosyl-(1->6)-alpha-D-glucopyranosyl beta-D-fructofuranoside; Melitose; Melitriose
Type Analytical Standard
Quantity 100 mg
Formula Weight 594.51
Percent Purity ≥99% (HPLC)
Grade Analytical Standard
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.