Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

D-(+)-Raffinose Pentahydrate, MP Biomedicals™

Catalog No. MP210279750 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
50 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP210279750 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MP210279750 Supplier MP Biomedicals, Inc Supplier No. 0210279750
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Composed of galactose, fructose and glucose. D-(+)-Raffinose Pentahydrate, MP Biomedicals is a trisaccharide and the most common alpha-galactosyl derivative of sucrose. Raffinose is hydrolysed to D-galactose and sucrose by D-galactosidase. This product is

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 17629-30-0
Molecular Formula C18H42O21
Molecular Weight (g/mol) 594.513
InChI Key BITMAWRCWSHCRW-ORBWWOJFSA-N
Synonym d-+-raffinosepentahydrate
PubChem CID 134129414
IUPAC Name (3S,4S,5R,6R)-2-[[(2R,3S,4S,5S)-6-[(2S,3S,4R,5R)-3,4-dihydroxy-2,5-bis(hydroxymethyl)oxolan-2-yl]oxy-3,4,5-trihydroxyoxan-2-yl]methoxy]-6-(hydroxymethyl)oxane-3,4,5-triol;pentahydrate
SMILES C(C1C(C(C(C(O1)OCC2C(C(C(C(O2)OC3(C(C(C(O3)CO)O)O)CO)O)O)O)O)O)O)O.O.O.O.O.O

Specifications

Melting Point 76°C to 86°C
Density 1.465g/mL (Lit.)
Quantity 50 g
Solubility Information Soluble in pyridine and water (1g/7mL).
Optical Rotation 101-107° (c=4, water)
Formula Weight 594.5
Percent Purity >98%
Physical Form Powder
Recommended Storage Store at Room Temperature (15-30°C).
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.