Learn More
Medchemexpress LLC D-ribofuranose-13C5 | 97.0% | 155.09 g·mol⁻1 | 13C5H10O5 | 1mg
SDP
Catalog No. 5000269512
Encompass_Preferred
D-Ribofuranose-13C5 (D-Ribose-13C5) is the 13C-labeled D-Ribofuranose (HY-113375) D-Ribofuranose (D-Ribose) is an endogenous metabolite present in Cerebrospinal Fluid that can be used for the research of Ribose 5 Phosphate Isomerase Deficiency and Medium Chain Acyl Co A Dehydrogenase Deficiency[1][2]
Catalog No. 50-002-69512 Supplier Medchemexpress LLC Supplier No. HYW7683331MG
D-Ribofuranose-13C5 (D-Ribose-13C5) is the 13C-labeled D-Ribofuranose (HY-113375) D-Ribofuranose (D-Ribose) is an endogenous metabolite present in Cerebrospinal Fluid that can be used for the research of Ribose 5 Phosphate Isomerase Deficiency and Medium Chain Acyl Co A Dehydrogenase Deficiency[1][2]