Learn More
Medchemexpress LLC D-ribofuranose-13C5 | 97.0% | 25mg
SDP
Catalog No. 5000270400
Encompass_Preferred
D-Ribofuranose-13C5 (D-Ribose-13C5) is the 13C-labeled D-Ribofuranose (HY-113375) D-Ribofuranose (D-Ribose) is an endogenous metabolite present in Cerebrospinal Fluid that can be used for the research of Ribose 5 Phosphate Isomerase Deficiency and Medium Chain Acyl Co A Dehydrogenase Deficiency[1][2]
Catalog No. 50-002-70400 Supplier Medchemexpress LLC Supplier No. HYW76833325MG
D-Ribofuranose-13C5 (D-Ribose-13C5) is the 13C-labeled D-Ribofuranose (HY-113375) D-Ribofuranose (D-Ribose) is an endogenous metabolite present in Cerebrospinal Fluid that can be used for the research of Ribose 5 Phosphate Isomerase Deficiency and Medium Chain Acyl Co A Dehydrogenase Deficiency[1][2]