Learn More
Cayman Chemical D-RIbonIc AcId -lacton 1g
SDP
Catalog No. 502918632
Encompass_Preferred
An oxidized derivative of ribose; has been used as a starting material in the synthesis of lasalocid, aristeromycine and (−)-neplanocin A, as well as an intermediate in the synthesis of ganglioside GM1 mimetic cholera toxin binder ligands
Catalog No. 50-291-8632 Supplier Cayman Chemical Supplier No. 392931 G
An oxidized derivative of ribose; has been used as a starting material in the synthesis of lasalocid, aristeromycine and (−)-neplanocin A, as well as an intermediate in the synthesis of ganglioside GM1 mimetic cholera toxin binder ligands