Learn More
Cayman Chemical D-(+)-Ribonic Acid γ-lactone, Purity: ≥98%, CAS: 5336-08-3, 5 g
SDP
Catalog No. 502916873
Encompass_Preferred
An oxidized derivative of ribose; has been used as a starting material in the synthesis of lasalocid, aristeromycine and (−)-neplanocin A, as well as an intermediate in the synthesis of ganglioside GM1 mimetic cholera toxin binder ligands
Catalog No. 50-291-6873 Supplier Cayman Chemical Supplier No. 392935 G
An oxidized derivative of ribose; has been used as a starting material in the synthesis of lasalocid, aristeromycine and (−)-neplanocin A, as well as an intermediate in the synthesis of ganglioside GM1 mimetic cholera toxin binder ligands