Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

D(+)-Ribonic acid γ-lactone, 96%, Thermo Scientific™

Catalog No. AC227830250
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC227830250 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC227830250 Supplier Thermo Scientific Chemicals Supplier No. 227830250

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5337-03-1
Molecular Formula C6H10O3
Molecular Weight (g/mol) 130.14
InChI Key AVPKHOTUOHDTLW-UHFFFAOYSA-N
Synonym tetrahydro-2h-pyran-4-carboxylic acid, tetrahydropyran-4-yl-carboxylic acid, tetrahydropyran-4-carboxylic acid, tetrahydro-pyran-4-carboxylic acid, 2h-pyran-4-carboxylic acid, tetrahydro, 4-carboxytetrahydro-2h-pyran, 3,4,5,6-tetrahydro-2h-pyran-4-carboxylic acid, 2h-3,4,5,6-tetrahydropyran-4-carboxylic acid, pubchem9730, acmc-2097ha
PubChem CID 219302
IUPAC Name oxane-4-carboxylic acid
SMILES C1COCCC1C(=O)O

Specifications

Melting Point 83.0°C to 85.0°C
Quantity 25 g
Specific Rotation Condition + 18.00 (20.00°C c=5, H2O)
Specific Rotation + 18.00
Solubility Information Solubility in water: soluble.
Formula Weight 148.12
Chemical Name or Material D(+)-Ribonic acid γ-lactone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.