Learn More
Medchemexpress LLC D-Ribonolactone | 5336-08-3 | ≥98.0% | C5H8O5 | 10 G
SDP
Catalog No. 5000433662
Encompass_Preferred
D-Ribonolactone is a sugar lactone and an inhibitor of β-galactosidase of Escherichia coli with a Ki of 26 mM. This product is intended for research and analytical applications.
  • Inhibits β-galactosidase of Escherichia coli
  • Suitable for research and analytical applications
  • Appears as a white to off-white solid
  • Purity of ≥98.0%
  • Specific storage conditions for powder and in solvent to maintain stability
Catalog No. 50-004-33662 Supplier Medchemexpress LLC Supplier No. HY7669110G
D-Ribonolactone is a sugar lactone and an inhibitor of β-galactosidase of Escherichia coli with a Ki of 26 mM. This product is intended for research and analytical applications.
  • Inhibits β-galactosidase of Escherichia coli
  • Suitable for research and analytical applications
  • Appears as a white to off-white solid
  • Purity of ≥98.0%
  • Specific storage conditions for powder and in solvent to maintain stability