Learn More
Medchemexpress LLC D-Ribonolactone | 5336-08-3 | MFCD00063241 | 98.0% | C5H8O5 | 1 ML
SDP
Catalog No. 5000434465
Encompass_Preferred
D-Ribonolactone is a sugar lactone and an endogenous metabolite, acting as an inhibitor of β-galactosidase in *Escherichia coli* with a Ki of 26 mM. Supplied as a white to off-white solid with a molecular weight of 148.11, this product is for research use only.
  • Inhibits β-galactosidase of *Escherichia coli* (Ki: 26 mM)
  • Endogenous metabolite
  • White to off-white solid with a molecular weight of 148.11
  • For research use only
  • Soluble in DMSO at 125 mg/mL (ultrasonication may be required)
  • Powder storage: -20°C for 3 years, 4°C for 2 years
  • Solution storage: -80°C for 6 months, -20°C for 1 month
  • Offers detailed in vivo dissolution protocols
Catalog No. 50-004-34465 Supplier Medchemexpress LLC Supplier No. HY7669110MM/1ML
D-Ribonolactone is a sugar lactone and an endogenous metabolite, acting as an inhibitor of β-galactosidase in *Escherichia coli* with a Ki of 26 mM. Supplied as a white to off-white solid with a molecular weight of 148.11, this product is for research use only.
  • Inhibits β-galactosidase of *Escherichia coli* (Ki: 26 mM)
  • Endogenous metabolite
  • White to off-white solid with a molecular weight of 148.11
  • For research use only
  • Soluble in DMSO at 125 mg/mL (ultrasonication may be required)
  • Powder storage: -20°C for 3 years, 4°C for 2 years
  • Solution storage: -80°C for 6 months, -20°C for 1 month
  • Offers detailed in vivo dissolution protocols