Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC D-ribose-1-13C | 70849-24-0 | MFCD00064325 | 99.9% | 151.12 g/mol | C4(13C)H10O5 | 50 MG
SDP

Catalog No. 5000301030
Encompass_Preferred

D-Ribose-13C is a carbon-13 labeled D-ribose used as a stable-isotope tracer for metabolic studies and analytical applications such as mass spectrometry and NMR. It provides high isotopic enrichment and high chemical purity to support accurate metabolic flux analysis and structural characterization.

  • 13C-labeled at carbon-1 for targeted isotope tracing.
  • High isotopic enrichment (99% 13C) for sensitive detection.
  • High chemical purity (HPLC 99.93%) to minimize impurities.
  • White to off-white solid suitable for analytical workflows.
  • Molecular weight 151.12 g/mol and formula C4(13C)H10O5.
  • Available in research-scale quantities (for example, 50 mg).

Catalog No. 50-003-01030 Supplier Medchemexpress LLC Supplier No. HYW018772S150MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

D-Ribose-13C is a carbon-13 labeled D-ribose used as a stable-isotope tracer for metabolic studies and analytical applications such as mass spectrometry and NMR. It provides high isotopic enrichment and high chemical purity to support accurate metabolic flux analysis and structural characterization.

  • 13C-labeled at carbon-1 for targeted isotope tracing.
  • High isotopic enrichment (99% 13C) for sensitive detection.
  • High chemical purity (HPLC 99.93%) to minimize impurities.
  • White to off-white solid suitable for analytical workflows.
  • Molecular weight 151.12 g/mol and formula C4(13C)H10O5.
  • Available in research-scale quantities (for example, 50 mg).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.