Learn More
Alkali Scientific D Ribose, 100 Grams
SDP
Catalog No. 5000660358
Encompass_Preferred
D Ribose, a high-purity sugar, is used primarily in biochemical research, including studies of metabolic pathways, energy production, and enzymatic reactions. This 100-gram bottle is ideal for experiments requiring a precise and consistent source of ribose. D-Ribose plays a critical role in the production of ATP, RNA, and DNA, making it essential for studies in cellular energy metabolism, nucleic acid synthesis, and cell signaling. It is also used in clinical research and the development of therapeutic agents targeting metabolic disorders. This product is ideal for laboratory use in molecular biology, biochemistry, and cellular research applications.
Catalog No. 50-006-60358 Supplier Alkali Scientific Supplier No. DRI001CP
D Ribose, a high-purity sugar, is used primarily in biochemical research, including studies of metabolic pathways, energy production, and enzymatic reactions. This 100-gram bottle is ideal for experiments requiring a precise and consistent source of ribose. D-Ribose plays a critical role in the production of ATP, RNA, and DNA, making it essential for studies in cellular energy metabolism, nucleic acid synthesis, and cell signaling. It is also used in clinical research and the development of therapeutic agents targeting metabolic disorders. This product is ideal for laboratory use in molecular biology, biochemistry, and cellular research applications.